Learn More
Foxx Life Sciences Vactrap™ G Vaccum Trap System (Vacuum Flask Alternative) for Aspiration and Vacuum Protection, 2 Liter PUREGRIP Glass Bottles Assembly with Secondary Container, Autoclavable, Inline Vent Filter, 1/EA
GREENER_CHOICE
Catalog No. NC1796900
Encompass_Preferred
Vactrap™ G (Glass) is the ultimate laboratory vacuum system nested neatly in a secondary container with a small footprint and incredible stability against tipping adhering to the strictest CDC recommendations for laboratory safety. The PUREGRIP glass media bottle with extra length EZFlow TPE tubing and tangle-free Polypropylene VersaCap ensures a continuous full vacuum rating. A 0.2μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump from waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration. Vactrap™ G combines ease-of-use with the highest temperature rating in the industry, 142°C.•40 CFR 264.175 compliant•3.3 Borosilicate Type 1 Glass•Bleach-Compatible•Autoclavable
Catalog No. NC1796900 Supplier Foxx Life Sciences Supplier No. 30R2001FLS
Vactrap™ G (Glass) is the ultimate laboratory vacuum system nested neatly in a secondary container with a small footprint and incredible stability against tipping adhering to the strictest CDC recommendations for laboratory safety. The PUREGRIP glass media bottle with extra length EZFlow TPE tubing and tangle-free Polypropylene VersaCap ensures a continuous full vacuum rating. A 0.2μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump from waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration. Vactrap™ G combines ease-of-use with the highest temperature rating in the industry, 142°C.•40 CFR 264.175 compliant•3.3 Borosilicate Type 1 Glass•Bleach-Compatible•Autoclavable