Learn More
Chemglass Life Sciences 2000mL Addition Funnel, Graduated, 24/40 Joint Size, 6mm PTFE Stpk, 525mm OAH
SDP
Catalog No. CG171208
Encompass_Preferred
2000mL Addition Funnel, Graduated, 24/40 Joint Size, 6mm PTFE Stopcock, 525mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171208 Supplier Chemglass Life Sciences Supplier No. CG171208
2000mL Addition Funnel, Graduated, 24/40 Joint Size, 6mm PTFE Stopcock, 525mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.