Learn More
Alkali Scientific 2L High Efficiency Erlenmeyer Flask Multifunctional Transfer Cap with TPE Tube (52cm 1/8" ID 1/4" OD) and a Sampling Nozzle, Vent Filter (0.22 μm Φ50mm), without Flask, Sterile, 1/pk, 4/cs
SDP
Catalog No. 503027413
Encompass_Preferred
Multifunctional Transfer Cap: For aseptic transfer of culture medium and cell culture solution. One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion. This cap meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121℃.There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume. Product Features High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterilized by E-beam, SAL=10^-6. Endotoxin-free, and no ingredients of animal origin. Individually packaged in sterile bag. Compatible with 2L high-efficient Erlenmeyer flasks. 
Catalog No. 50-302-7413 Supplier Alkali Scientific Supplier No. EFC409
Multifunctional Transfer Cap: For aseptic transfer of culture medium and cell culture solution. One end of the pipeline is connected to the MPC series connector, which increases the convenience and safety in use. The male connector and the sealing cap are tightly connected to prevent accidental disconnection, and the connector can be rotated to reduce pipeline distortion. This cap meets the USP Class 6 Standard, with high transparency, has strong impact resistance, oxidation resistance and can withstand a high temperature up to 121℃.There’s a scale made by injection molding on the flask body to facilitate observing the filled liquid volume. Product Features High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance. Sterilized by E-beam, SAL=10^-6. Endotoxin-free, and no ingredients of animal origin. Individually packaged in sterile bag. Compatible with 2L high-efficient Erlenmeyer flasks. 