Learn More
Alkali Scientific 2L High Efficiency Erlenmeyer
SDP
Catalog No. 503027414
Encompass_Preferred
Multifunctional Transfer Cap For aseptic transfer of culture medium and cell culture solution One end of the pipeline is connected to the MPC series connector which increases the convenience and safety in use The male connector and the sealing cap are tightly connected to prevent accidental disconnection and the connector can be rotated to reduce pipeline distortion This cap meets the USP Class 6 Standard with high transparency has strong impact resistance oxidation resistance and can withstand a high temperature up to 121 Theres a scale made by injection molding on the flask body to facilitate observing the filled liquid volume Product Features High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance Sterilized by E-beam SAL10-6 Endotoxin-free and no ingredients of animal origin Individually packaged in sterile bag Compatible with 2L high-efficient Erlenmeyer flasks 50cm long external TPE transfer tube 1/4 in Internal diameter 7/16
Catalog No. 50-302-7414 Supplier Alkali Scientific Supplier No. EFC410
Multifunctional Transfer Cap For aseptic transfer of culture medium and cell culture solution One end of the pipeline is connected to the MPC series connector which increases the convenience and safety in use The male connector and the sealing cap are tightly connected to prevent accidental disconnection and the connector can be rotated to reduce pipeline distortion This cap meets the USP Class 6 Standard with high transparency has strong impact resistance oxidation resistance and can withstand a high temperature up to 121 Theres a scale made by injection molding on the flask body to facilitate observing the filled liquid volume Product Features High-quality materials and smooth inner wall of the tubing provide an excellent transfer performance Sterilized by E-beam SAL10-6 Endotoxin-free and no ingredients of animal origin Individually packaged in sterile bag Compatible with 2L high-efficient Erlenmeyer flasks 50cm long external TPE transfer tube 1/4 in Internal diameter 7/16