Learn More
Ace Glass, Inc. 2000mL jacketed pressure reactor, complete with #11 Ace-Thred inlet/outlets, 100mm Ace-Thred 4 neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
SDP
Catalog No. 50972817
Encompass
2000mL jacketed pressure reactor, complete with #11 Ace-Thred inlet/outlets, 100mm Ace-Thred 4 neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
Catalog No. 50-972-817 Supplier Ace Glass, Inc. Supplier No. 6386520
Add to Cart
Add to Cart
Request a Quote
2000mL jacketed pressure reactor, complete with #11 Ace-Thred inlet/outlets, 100mm Ace-Thred 4 neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings