Learn More
Ace Glass, Inc. 2000mL separatory funnel with locking stopper.
SDP
Catalog No. 50974702
Encompass_Preferred
2000ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-702 Supplier Ace Glass, Inc. Supplier No. 723165
Add to Cart
Add to Cart
2000ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.