Learn More
Andwin Scientific 2L MEDI VAC SUCTION CANISTERS
SDP
Catalog No. NC9516182
Encompass_Preferred
The Guardian Canisters are designed with rigid blue lids with an automatic shutoff valve to eliminate the risk of cross contamination of regulators and wall vacuum outlets. The locking lid promotes the correct way of discarding of infectious liquid medical waste and improves user safety. This product has a volume capacity of 2 liters.
This item is not returnable. View return policy
Catalog No. NC9516182 Supplier Andwin Scientific Supplier No. 65651220
The Guardian Canisters are designed with rigid blue lids with an automatic shutoff valve to eliminate the risk of cross contamination of regulators and wall vacuum outlets. The locking lid promotes the correct way of discarding of infectious liquid medical waste and improves user safety. This product has a volume capacity of 2 liters.