Learn More
NEST SCIENTIFIC INC 2L PC Conical Erlenmeyer Flasks, Flat Base, Vent Filter Cap, Sterile, 6/cs
Catalog No. NC2887694
Encompass_Preferred
Nest Scientific PC shaker flasks are ideal for growing suspended cell lines. Molded from USP grade VI virgin PC which may be autoclaved up to 6 times. Flasks feature a non-wettable 0.22um PTFE membrane for sterile gas exchange. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC2887694 Supplier NEST SCIENTIFIC INC Supplier No. 785011
Nest Scientific PC shaker flasks are ideal for growing suspended cell lines. Molded from USP grade VI virgin PC which may be autoclaved up to 6 times. Flasks feature a non-wettable 0.22um PTFE membrane for sterile gas exchange. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.