Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 3-Neck, 29/42 CN - 24/40 SN's Angled 20°
SDP
Catalog No. CG152451
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40
Catalog No. CG152451 Supplier Chemglass Life Sciences Supplier No. CG152451
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿.Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40