Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 3-Neck, 29/42 CN - 29/42 SN's, Vertical, Half Jacketed
SDP
Catalog No. CG153709
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Vertical, Half Jacketed. Round bottom, HEAVY WALL, three-neck flask with standard taper outer joints on the center neck and vertical side necks. Lower half of the flask is jacketed with two hose connections for circulation of heating or cooling liquid.
Catalog No. CG153709 Supplier Chemglass Life Sciences Supplier No. CG153709
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Vertical, Half Jacketed. Round bottom, HEAVY WALL, three-neck flask with standard taper outer joints on the center neck and vertical side necks. Lower half of the flask is jacketed with two hose connections for circulation of heating or cooling liquid.