Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 4-Neck, 24/40 CN - 24/40 SN's, # 7 Chem-Thread Side Neck
SDP
Catalog No. CG153404
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 24/40 - 24/40, # 7 Chem-Thread Side Neck, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints and a # 7 Chem-Thread for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.
Catalog No. CG153404 Supplier Chemglass Life Sciences Supplier No. CG153404
Flask, Round Bottom, 2000mL, Heavy Wall, 24/40 - 24/40, # 7 Chem-Thread Side Neck, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints and a # 7 Chem-Thread for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.