Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 4-Neck, 29/26 CN - 29/26 SN's, 10/30 Thermometer Joint
SDP
Catalog No. CG153311
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 29/26 - 29/26, 10/30 Thermometer Joint, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints on the center neck and vertical side necks. The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in the clamping of the flask.
Catalog No. CG153311 Supplier Chemglass Life Sciences Supplier No. CG153311
Flask, Round Bottom, 2000mL, Heavy Wall, 29/26 - 29/26, 10/30 Thermometer Joint, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints on the center neck and vertical side necks. The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in the clamping of the flask.