Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 4-Neck, 29/42 CN - 24/40 SN's, 10/30 Thermometer Joint
SDP
Catalog No. CG153312
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 24/40, 10/30 Thermometer Joint, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints on the center neck and vertical side necks. The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer.
Catalog No. CG153312 Supplier Chemglass Life Sciences Supplier No. CG153312
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 24/40, 10/30 Thermometer Joint, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints on the center neck and vertical side necks. The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer.