Learn More
Chemglass Life Sciences Flask, Round Bottom, 2000mL, Heavy Wall, 3-Neck, 29/42 CN - 29/42 SNs, Vertical
SDP
Catalog No. CG152514
Encompass_Preferred
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Vertical. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and side necks. Indentations increase agitation of the contents of the flask during stirring. Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum
Catalog No. CG152514 Supplier Chemglass Life Sciences Supplier No. CG152514
Flask, Round Bottom, 2000mL, Heavy Wall, 29/42 - 29/42, 3-Neck, Vertical. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center and side necks. Indentations increase agitation of the contents of the flask during stirring. Please Note: Morton Style Reactors/Flasks Not Suitable For Vacuum