Learn More
Chemglass Life Sciences 2000mL Reservoir, Chromatography, 35/20 Lower Ball
SDP
Catalog No. CG120035
Encompass_Preferred
2000mL Reservoir, Chromatography, 35/20 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.
Catalog No. CG120035 Supplier Chemglass Life Sciences Supplier No. CG120035
2000mL Reservoir, Chromatography, 35/20 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.