Learn More
Alkali Scientific Alkali Scientific™ 2L Roller Bottles (2000mL), 850cm² Surface Area, Individually Wrapped, Sterile, Printed Graduations, Easy Grip Cap, 12/Cs, Non-Treated, Vent Cap (0.22µm hydrophobic membrane)
SDP
Catalog No. 503028419
Encompass_Preferred
Our 2000mL Easy Grip Roller Bottle is an essential tool for tissue culture applications. Designed with convenience in mind, this roller bottle features printed graduations for easy volume estimation. The inclusion of a vented cap ensures optimal gas exchange while maintaining a sterile environment. Ideal for research and laboratory settings, this roller bottle facilitates the growth and maintenance of cell cultures with precision Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0.22µm hydrophobic membrane
Catalog No. 50-302-8419 Supplier Alkali Scientific Supplier No. VNT2100V
Our 2000mL Easy Grip Roller Bottle is an essential tool for tissue culture applications. Designed with convenience in mind, this roller bottle features printed graduations for easy volume estimation. The inclusion of a vented cap ensures optimal gas exchange while maintaining a sterile environment. Ideal for research and laboratory settings, this roller bottle facilitates the growth and maintenance of cell cultures with precision Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0.22µm hydrophobic membrane