Learn More
Chemglass Life Sciences 2000mL Solvent Reservoir, Plastic Coated, Chromatography
SDP
Catalog No. CG116604
Encompass_Preferred
2000mL Solvent Reservoir, Plastic Coated, Chromatography. Reservoirs used for storing high performance, low pressure liquid chromatography mobile phase. Complete reservoir system includes a plastic coated graduated bottle, a standard cap assembly, & solvent & sparging filters. Cap assembly fits GL45 threaded bottles & has a PTFE bulkhead w/ three 1/4in- 28 threaded ports to accommodate fittings for 1/8in O.D. tubing. Fittings are supplied w/ the cap assemblies. There are no threads or fittings, just insert the 1/8in O.D. tubing through the holes in the Tefzel bulkhead. The solvent & sparging filters have a 2m stainless steel replaceable filter cup with a ferrule & nut for 1/8in O.D. tubing. A 10m filter assembly is offered as an option. The solvent reservoir stand positions the filters for maximum uptake of the mobile phase. An economical solvent & sparging filter is available w/ all PEEK construction & a 2m filter. PTFE (FEP) tubing is sold separately.
Catalog No. CG116604 Supplier Chemglass Life Sciences Supplier No. CG116604
2000mL Solvent Reservoir, Plastic Coated, Chromatography. Reservoirs used for storing high performance, low pressure liquid chromatography mobile phase. Complete reservoir system includes a plastic coated graduated bottle, a standard cap assembly, & solvent & sparging filters. Cap assembly fits GL45 threaded bottles & has a PTFE bulkhead w/ three 1/4in- 28 threaded ports to accommodate fittings for 1/8in O.D. tubing. Fittings are supplied w/ the cap assemblies. There are no threads or fittings, just insert the 1/8in O.D. tubing through the holes in the Tefzel bulkhead. The solvent & sparging filters have a 2m stainless steel replaceable filter cup with a ferrule & nut for 1/8in O.D. tubing. A 10m filter assembly is offered as an option. The solvent reservoir stand positions the filters for maximum uptake of the mobile phase. An economical solvent & sparging filter is available w/ all PEEK construction & a 2m filter. PTFE (FEP) tubing is sold separately.