Learn More
Alkali Scientific 2L Square Media Storage Bottle
SDP
Catalog No. 503026374
Encompass_Preferred
The 2L Square Media Storage Bottle is a high-quality reagent storage solution designed for laboratory applications Made from durable polycarbonate (PC) it offers excellent clarity impact resistance and chemical durability ensuring safe and efficient liquid storage Durable Polycarbonate Construction Provides high transparency strong mechanical strength and superior impact resistance Sterile and Safe Non-cytotoxic non-pyrogenic and free of animal-derived components making it suitable for sensitive laboratory environments Chemical and Environmental Resistance Tolerates low temperatures UV exposure and cracking ensuring long-term usability Space-Efficient Square Design Enhances storage efficiency and stability making it easy to handle and stack Smooth Inner Surface Minimizes residue buildup facilitating easy cleaning and reducing contamination risks Leak-Proof Cap Specially designed cap formed through one-shot molding ensures a secure and tight seal without an inner gasket Er
Catalog No. 50-302-6374 Supplier Alkali Scientific Supplier No. VB2000PC
The 2L Square Media Storage Bottle is a high-quality reagent storage solution designed for laboratory applications Made from durable polycarbonate (PC) it offers excellent clarity impact resistance and chemical durability ensuring safe and efficient liquid storage Durable Polycarbonate Construction Provides high transparency strong mechanical strength and superior impact resistance Sterile and Safe Non-cytotoxic non-pyrogenic and free of animal-derived components making it suitable for sensitive laboratory environments Chemical and Environmental Resistance Tolerates low temperatures UV exposure and cracking ensuring long-term usability Space-Efficient Square Design Enhances storage efficiency and stability making it easy to handle and stack Smooth Inner Surface Minimizes residue buildup facilitating easy cleaning and reducing contamination risks Leak-Proof Cap Specially designed cap formed through one-shot molding ensures a secure and tight seal without an inner gasket Er