Learn More
Ace Glass, Inc. 2L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 3/8 id tubing
SDP
Catalog No. 5000810369
Encompass_Preferred
2L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 3/8 id tubing
Catalog No. 50-008-10369 Supplier Ace Glass, Inc. Supplier No. 538905
2L StreamFlow Bottle with Hose Connection includes expanded PTFE membrane vented cap GL-45 thread hose connection for 3/8 id tubing