Learn More
Chemglass Life Sciences 2L Unjacketed Bio Reactor Vessel
SDP
Catalog No. 501173062
Encompass
2L Unjacketed Bio Reactor Vessel. Vessels are constructed of heavy wall tubing and are graduated in 200mL subdivisions with numbers every 1000mL. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-117-3062 Supplier Chemglass Life Sciences Supplier No. CLS137901
2L Unjacketed Bio Reactor Vessel. Vessels are constructed of heavy wall tubing and are graduated in 200mL subdivisions with numbers every 1000mL. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.