Learn More
Medicus Health Mini Water Bath, 2 Liter
SDP
Catalog No. NC3589286
Encompass
• Mini 2 Liter water bath is 6.5" wide and is great for thawing fresh frozen plasma or cryoprecipitate and is easy to clean• Thawing fresh frozen plasma or cryoprecipitate can really mess up your day if you get a cracked or broken bag in your water bath
• Lab water bath is small enough to be poured down the sink by hand and be disinfected in mere minutes to get back to your next task in no time
• Stainless steel bowl is smooth and there are no recirculators that need to get rinsed out
• Compact 2-liter mini water bath can fit at any workstation and features easy to use controls that allow the user to digitally select and monitor temperature
• Digital accuracy and convenience
• Quik-CAL™ user calibration function allows the end user to easily calibrate the unit for accurate temp readouts
• Dimensions: 6.5" W x 8" D x 6" H
Catalog No. NC3589286 Supplier Medicus Health Supplier No. 5462M2
• Mini 2 Liter water bath is 6.5" wide and is great for thawing fresh frozen plasma or cryoprecipitate and is easy to clean• Thawing fresh frozen plasma or cryoprecipitate can really mess up your day if you get a cracked or broken bag in your water bath
• Lab water bath is small enough to be poured down the sink by hand and be disinfected in mere minutes to get back to your next task in no time
• Stainless steel bowl is smooth and there are no recirculators that need to get rinsed out
• Compact 2-liter mini water bath can fit at any workstation and features easy to use controls that allow the user to digitally select and monitor temperature
• Digital accuracy and convenience
• Quik-CAL™ user calibration function allows the end user to easily calibrate the unit for accurate temp readouts
• Dimensions: 6.5" W x 8" D x 6" H