Learn More
Electron Microscopy Sciences Lyophilization Two Leg Stoppers for 7mm ID, 13 mm O.D.
SDP
Catalog No. 501898952
Encompass_Preferred
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 7 mm
O.D.: 13 mm
Material: Gray chlorobutyl/50
Catalog No. 50-189-8952 Supplier Electron Microscopy Sciences Supplier No. 6032607
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 7 mm
O.D.: 13 mm
Material: Gray chlorobutyl/50