Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Calcium Chloride (2M), 100 mL
SDP

Catalog No. 507517642
Encompass_Preferred

Calcium Chloride (2M) is useful in the preparation of competent E.coli and transformation of bacteria.

• Preparation of competent E. coli
• Transformation of bacteria
• 2 Molar concentration
• Filtered through a 0.22 µm membrane
• DNase, RNase, and Protease tested

Applications: Calcium chloride is a commonly used reagent in biochemistry. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7642 Supplier Quality Biological Inc Supplier No. 351130721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcium Chloride (2M) is useful in the preparation of competent E.coli and transformation of bacteria.

• Preparation of competent E. coli
• Transformation of bacteria
• 2 Molar concentration
• Filtered through a 0.22 µm membrane
• DNase, RNase, and Protease tested

Applications: Calcium chloride is a commonly used reagent in biochemistry. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.