Learn More
Teknova 2M Imidazole, pH 8.0. 1000mL, Sterile.
SDP
Supplier:  Teknova I7080
Encompass
Imidazole is a small organic compound that is used as a buffer in protein purification to prevent changes in pH, helping to maintain stability of products. It is also used as an antifungal agent and has a wide range of other uses in the food, beverage, and pharmaceutical industries.
Catalog No. NC0673205