Learn More
Teknova 2M Potassium Chloride Solution. 1000 mL, Sterile.
SDP
Catalog No. 50842961
Encompass
Potassium chloride solution, also known as KCl, is a salt that has a wide range of applications in molecular biology, protein chemistry and biochemical research. It is a key component of many biochemical reactions, including the synthesis of proteins and DNA. In addition, KCl is a popular reagent for the purification of proteins and nucleic acids.
This item is not returnable. View return policy
Catalog No. 50-842-961 Supplier Teknova Supplier No. P0335
Potassium chloride solution, also known as KCl, is a salt that has a wide range of applications in molecular biology, protein chemistry and biochemical research. It is a key component of many biochemical reactions, including the synthesis of proteins and DNA. In addition, KCl is a popular reagent for the purification of proteins and nucleic acids.