Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Magnesium Chloride (2M), 100 mL
SDP

Catalog No. 507517307
Encompass_Preferred

Magnesium is a typical ionic halide, and is very soluble in water. Magnesium is an essential co-factor in many enzymes, including DNAse, some restriction enzymes, and Ribonuclease H.

Used with SOC medium

Magnesium has a variety of biological roles in enzymology, cell membrane/wall structure, muscle cell physiology, and nucleic acid structure. Can be added to SOB and SOC medium at a concentration of 20 mM (10 mL 2M MgCl2 per liter).

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7307 Supplier Quality Biological Inc Supplier No. 340034721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnesium is a typical ionic halide, and is very soluble in water. Magnesium is an essential co-factor in many enzymes, including DNAse, some restriction enzymes, and Ribonuclease H.

Used with SOC medium

Magnesium has a variety of biological roles in enzymology, cell membrane/wall structure, muscle cell physiology, and nucleic acid structure. Can be added to SOB and SOC medium at a concentration of 20 mM (10 mL 2M MgCl2 per liter).

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.