Learn More
Teknova 2M MOPS Buffer, pH 7.4. 500mL, Sterile 8 pack.
SDP
Catalog No. 502387666
Encompass
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.
Catalog No. 50-238-7666 Supplier Teknova Supplier No. M84058PK
Add to Cart
Add to Cart
MOPS (3-(N-morpholino)propanesulfonic acid) is a Good’s buffer and has a pKa of 7.01, making it an ideal buffer for many biological systems at near-neutral pH. MOPS can be used in cell culture media and biopharma buffer formulations as well as in the purification of antibodies, peptides, proteins and blood components. It is nontoxic to cell culture lines.