Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Tris Base Solutions
GREENER_CHOICE

Catalog No. A65690BA
Change view
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
Concentration:
1 M
2 M
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
A65690BA 5 L 2 M
A65690BB 20 L 2 M
A65690BC 50 L 2 M
A65690BD 100 L 2 M
A65690BE 200 L 2 M
A65784BA 5 L 1 M
A65784BB 20 L 1 M
A65784BC 50 L 1 M
A65784BD 100 L 1 M
A65784BE 200 L 1 M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. A65690BA Supplier Gibco™ Supplier No. A65690BA
Add to Cart
Add to Cart

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-86-1
Concentration 2 M
For Use With (Application) Bioproduction
For Use With (Equipment) ILC/ILD System
pH 10.0 to 12.0
Physical Form Solution
Quantity 5 L
Solution Type Buffer
Packaging Bioprocess Container in Secondary Packaging
Manufacturing Quality CGMP
Molecular Weight (g/mol) Raw Material: 121.14
Percent Purity Not Available
Grade EP/USP
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.