Learn More
Brainxell Inc Microglia (Human iPSC-Derived, Female Line, 2M)
SDP
Catalog No. NC2309450
Encompass
Microglia derived from iPSC line WC-32, female. Contents: 1 vial of ≥2x10^6 viable cryopreserved cells. Marker Expression: Confirmed expression of microglia surface markers including: CD45, CD11b, CX3CR1, P2RY12, and TMEM119. Immunocytochemistry shortly after plating microglia (shown here with a DAPI stain) reveals prominent expression of TREM2 (green) and IBA1 (red). Function: Human microglia demonstrate phagocytosis and can be stimulated to secrete a variety of cytokines. iPSC derived microglia are immunologically naive. Related diseases: Autism, Epilepsy, Multiple Sclerosis, Alzheimer's & Huntington's Disease.
Catalog No. NC2309450 Supplier Brainxell Inc Supplier No. BX0900322M
Microglia derived from iPSC line WC-32, female. Contents: 1 vial of ≥2x10^6 viable cryopreserved cells. Marker Expression: Confirmed expression of microglia surface markers including: CD45, CD11b, CX3CR1, P2RY12, and TMEM119. Immunocytochemistry shortly after plating microglia (shown here with a DAPI stain) reveals prominent expression of TREM2 (green) and IBA1 (red). Function: Human microglia demonstrate phagocytosis and can be stimulated to secrete a variety of cytokines. iPSC derived microglia are immunologically naive. Related diseases: Autism, Epilepsy, Multiple Sclerosis, Alzheimer's & Huntington's Disease.