Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag bioMIXcontrol S AC/DC1EA
Catalog No. 502908607
Encompass
.nbspEfficient control unit for 2mag bioMIXdrive series developed for the hard daily lab use and uncompromising warming-free continuous operation.nbspControl unit for the connection of 1 stirring drive bioMIXdrive 1 2 3 4 or 6 with 6 watts/stirring point without any own-generated heating effect extended speed range from 5 to 250 rpm jerk-free stirring even and smooth stirring also at slow speeds.nbspPower setting (10-step) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation e.g. in CO2-incubators.nbspIdentical to bioMIXcontrol but with stackable housing and vertical control panel.nbspDigital display for speed- and power setting SoftStart for reliable and safe accelerating for stirring bar QuickSet for quick setting of start- and maximum speed storage of last working status when turning off firm stainless steel housing and easy to clean
Catalog No. 50-290-8607 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7645581EA
.nbspEfficient control unit for 2mag bioMIXdrive series developed for the hard daily lab use and uncompromising warming-free continuous operation.nbspControl unit for the connection of 1 stirring drive bioMIXdrive 1 2 3 4 or 6 with 6 watts/stirring point without any own-generated heating effect extended speed range from 5 to 250 rpm jerk-free stirring even and smooth stirring also at slow speeds.nbspPower setting (10-step) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation e.g. in CO2-incubators.nbspIdentical to bioMIXcontrol but with stackable housing and vertical control panel.nbspDigital display for speed- and power setting SoftStart for reliable and safe accelerating for stirring bar QuickSet for quick setting of start- and maximum speed storage of last working status when turning off firm stainless steel housing and easy to clean