Learn More
2Mag USA 2mag MIX 1 XL Stirrer (with power supply)
SDP
Catalog No. NC1781568
Encompass
Direct controlled, single point stirrer; digital display; for large volume and industrial applications; stirring volume up to 40 liters; stirring speed range from 70 to 2,000 rpm; adjustable power setting with up to 20 watts of stirring power; includes power supply.
Catalog No. NC1781568 Supplier 2Mag USA Supplier No. MF 30201
Direct controlled, single point stirrer; digital display; for large volume and industrial applications; stirring volume up to 40 liters; stirring speed range from 70 to 2,000 rpm; adjustable power setting with up to 20 watts of stirring power; includes power supply.