Learn More
2Mag USA 2MAG MIX 15 ECO STIRRER/PWR SP
SDP
Catalog No. NC1650488
Encompass
Direct controlled, multi point stirrer with 15 synchronized stirring positions; digital display; stirring volume up to ­­1,500 milliliters; stirring speed range from 120 to 1,200 rpm; up to 10 watts of stirring power; includes power supply.
Catalog No. NC1650488 Supplier 2Mag USA Supplier No. MF30115
Direct controlled, multi point stirrer with 15 synchronized stirring positions; digital display; stirring volume up to ­­1,500 milliliters; stirring speed range from 120 to 1,200 rpm; up to 10 watts of stirring power; includes power supply.