Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIX 15 multi position1EA
Catalog No. 502908542
Encompass
Ultra-flat magnetic stirrer with 15 stirring positions up to 3000 mL stirring volume suitable for 15 x 250 mL beaker glasses.Powerful 15 stirring positions 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept extremely wide speed range of 100 to 2000 rpm 100% synchronized speed jerk-free stirring even at low speeds 4-step power settings of the stirrer high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer..nbspMaintenance-free.nbspSynchronized stirring of 15 x 250 mL beakers.nbspSoftStar setting for reliable jerk-free centering of stirring bar.nbspClear digital display.nbspAutomatic memory saves last settings
Catalog No. 50-290-8542 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6786191EA
Add to Cart
Add to Cart
Request a Quote
Ultra-flat magnetic stirrer with 15 stirring positions up to 3000 mL stirring volume suitable for 15 x 250 mL beaker glasses.Powerful 15 stirring positions 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept extremely wide speed range of 100 to 2000 rpm 100% synchronized speed jerk-free stirring even at low speeds 4-step power settings of the stirrer high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer..nbspMaintenance-free.nbspSynchronized stirring of 15 x 250 mL beakers.nbspSoftStar setting for reliable jerk-free centering of stirring bar.nbspClear digital display.nbspAutomatic memory saves last settings