Learn More
Sigma Aldrich Fine Chemicals Biosciences 2MAG MIX 4 MS multiple 4 p1EA
Catalog No. 502908540
Encompass
Ultra-flat magnetic stirrer with 4 individually controlled stirring positions suitable for 4 x 2000 ml Erlenmeyer flasks.Powerful 4 stirring positions 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive conceptpower reserves up to 3000 ml per stirring point individualand independent speed setting for each stirring drive(Master-Slave-Technology) also ON/OFF functionsynchronous speed mode for all positions extremelywide speed range of 100 to 2000 rpm jerk-free stirringeven at low speeds.
Catalog No. 50-290-8540 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423241EA
Add to Cart
Add to Cart
Request a Quote
Ultra-flat magnetic stirrer with 4 individually controlled stirring positions suitable for 4 x 2000 ml Erlenmeyer flasks.Powerful 4 stirring positions 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive conceptpower reserves up to 3000 ml per stirring point individualand independent speed setting for each stirring drive(Master-Slave-Technology) also ON/OFF functionsynchronous speed mode for all positions extremelywide speed range of 100 to 2000 rpm jerk-free stirringeven at low speeds.