Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIX 6 multi position1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6786431EA
Encompass
Ultra-flat magnetic stirrer with 6 stirring positions up to 6000 mL stirring volume suitable for 6 x 1000 mL flasks.Powerful 6 stirring positions 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept extremely wide speed range of 100 to 2000 rpm 100% synchronized speed jerk-free stirring even at low speeds 4-step power settings of the stirrer high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer..nbspMaintenance-free.nbspSynchronized stirring of 6 x 1000 mL flasks.nbspSoftStar setting for reliable jerk-free centering of stirring bar.nbspClear digital display.nbspAutomatic memory saves last settings
Catalog No. 50-290-8519