Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIX 6 multi position1EA
Catalog No. 502908506
Encompass
Ultra-flat magnetic stirrer with 6 stirring positions up to 6 000 mL stirring volume suitable for 6 x 1 000 mL flasks.Powerful 6 stirring positions 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept extremely wide speed range of 100 to 2 000 rpm 100% synchronized speed jerk-free stirring even at low speeds 4-step power settings of the stirrer high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.Maintenance-freeSynchronized stirring of 6 x 1 000 mL flasksSoftStar setting for reliable jerk-free centering of stirring barClear digital displayAutomatic memory saves last settings
Catalog No. 50-290-8506 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6786351EA
Ultra-flat magnetic stirrer with 6 stirring positions up to 6 000 mL stirring volume suitable for 6 x 1 000 mL flasks.Powerful 6 stirring positions 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept extremely wide speed range of 100 to 2 000 rpm 100% synchronized speed jerk-free stirring even at low speeds 4-step power settings of the stirrer high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.Maintenance-freeSynchronized stirring of 6 x 1 000 mL flasksSoftStar setting for reliable jerk-free centering of stirring barClear digital displayAutomatic memory saves last settings