Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIXcontrol 20 control1EA
Catalog No. 502908509
Encompass
Control unit with 20 watts power speed range from 100 up to 1 600 rpm jerk-free stirring also at low speeds power setting (4-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives.Clear digital display for speed- and power settings SoftStart for reliable catching/centering and safe accelerating of the stirring bar. QuickSet for quick setting of start- and maximum speed automatic memory function to save the last settings resistant stainless steel housing easy to clean.
Catalog No. 50-290-8509 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5698601EA
Control unit with 20 watts power speed range from 100 up to 1 600 rpm jerk-free stirring also at low speeds power setting (4-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives.Clear digital display for speed- and power settings SoftStart for reliable catching/centering and safe accelerating of the stirring bar. QuickSet for quick setting of start- and maximum speed automatic memory function to save the last settings resistant stainless steel housing easy to clean.