Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIXcontrol eco contro1EA
Catalog No. 502908548
Encompass
Control unit for small to medium volumes and the connection of one stirring dirve MIXdrive 10W stirring power speed range from 100 to 1 200 rpm SoftStart for reliable catching/centering and safe accelerating of the stirring bar.Interfaces 0-10 volts 4-20 mA RS232 USB Ethernet.
Catalog No. 50-290-8548 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5698791EA
Control unit for small to medium volumes and the connection of one stirring dirve MIXdrive 10W stirring power speed range from 100 to 1 200 rpm SoftStart for reliable catching/centering and safe accelerating of the stirring bar.Interfaces 0-10 volts 4-20 mA RS232 USB Ethernet.