Learn More
Sigma Aldrich Fine Chemicals Biosciences 2MAG MIXdrive 1 single poi1EA
Catalog No. 502908580
Encompass
Ultra flat wear-free magnetic stirrer with 1 stirring position suitable for vessels up to 10 000 ml.1 stirring position 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerfulmixing of SOLUTIONons jerk-free stirring even at low speeds.Fully encapsulated and hermetically sealed stainless steelhousing IP68 water- dust- and germproof extremelyrobust construction and performance easy to clean andmaximum resistance submersible in water- or oil baths suitable for the use in CO2-incubators and ovens.
Catalog No. 50-290-8580 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423321EA
Ultra flat wear-free magnetic stirrer with 1 stirring position suitable for vessels up to 10 000 ml.1 stirring position 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerfulmixing of SOLUTIONons jerk-free stirring even at low speeds.Fully encapsulated and hermetically sealed stainless steelhousing IP68 water- dust- and germproof extremelyrobust construction and performance easy to clean andmaximum resistance submersible in water- or oil baths suitable for the use in CO2-incubators and ovens.