Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIXdrive 15 multi pos1EA
Catalog No. 502908573
Encompass
100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions 100% synchronized speeds jerk-free stirring even at low speeds.Clear digital display for settings of stirrer speed and power SoftStart procedure for reliable catching/centering and safe acceleration of the stirring bar QuickSet for quick setting of start- and maximum speed Automatic memory function to save the last settings outstanding long-life cycle of the hermetically sealed drive system resistant stainless steel housing for easy and fast cleaning.Stirring point distance 65 mm.Fully encapsulated and hermetically sealed stainless steel housing submersible in water-baths suitable for the use in CO2-incubators and ovens.Control unit sold separately.
Catalog No. 50-290-8573 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6935101EA
100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions 100% synchronized speeds jerk-free stirring even at low speeds.Clear digital display for settings of stirrer speed and power SoftStart procedure for reliable catching/centering and safe acceleration of the stirring bar QuickSet for quick setting of start- and maximum speed Automatic memory function to save the last settings outstanding long-life cycle of the hermetically sealed drive system resistant stainless steel housing for easy and fast cleaning.Stirring point distance 65 mm.Fully encapsulated and hermetically sealed stainless steel housing submersible in water-baths suitable for the use in CO2-incubators and ovens.Control unit sold separately.