Learn More
Sigma Aldrich Fine Chemicals Biosciences 2MAG MIXdrive 24 MTP1EA
Catalog No. 502909003
Encompass
Ultra flat wear-free magnetic stirrer with 6/12/24/96 stirring positions suitable for microtiter plates.All stirring positions 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions 100 percent synchronized speeds jerk-free stirring even at low speeds.Fully encapsulated and hermetically sealed stainless steelhousing IP68 water- dust- and germproof extremely robustconstruction and performance easy to clean and maximumresistance submersible in water- resp. oil baths up to 50 C.
Catalog No. 50-290-9003 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423711EA
Ultra flat wear-free magnetic stirrer with 6/12/24/96 stirring positions suitable for microtiter plates.All stirring positions 100 percent maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions 100 percent synchronized speeds jerk-free stirring even at low speeds.Fully encapsulated and hermetically sealed stainless steelhousing IP68 water- dust- and germproof extremely robustconstruction and performance easy to clean and maximumresistance submersible in water- resp. oil baths up to 50 C.