Learn More
Sigma Aldrich Fine Chemicals Biosciences 2mag MIXdrive 6 multi posi1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6935021EA
Encompass
Clear digital display for settings of stirrer speed and power SoftStart procedure for reliable catching/centering and safe acceleration of the stirring bar QuickSet for quick setting of start- and maximum speed Automatic memory function to save the last settings outstanding long-life cycle of the hermetically sealed drive system resistant stainless steel housing for easy and fast cleaning.100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of SOLUTIONons 100% synchronized speeds jerk-free stirring even at low speeds.Ultra flat wear-free magnetic stirrer with 6 stirring positions suitable for 6 x 250 mL beaker glasses.Stirring point distance 130 mm.Fully encapsulated and hermetically sealed stainless steel housing. Use with external control unit. Required but not supplied - Z569836 with EU and UK plugs or Z569879 with US plug
Catalog No. 50-290-8599