Learn More
AdipoGen 2MeO-4-coumarinylacetic acid
SDP
Supplier:  AdipoGen CDXD0034G001
Encompass_Preferred
Chemical. CAS 88404-26-6. Formula C13H12O6. MW 264.23. Synthetic. 6,7-Dimethoxy-4-coumarinyl acetic is a fluorescent molecule which may be used in fluorescent applications that use coumarinyl acetic acids.
Catalog No. 50-256-1299