Learn More
Cayman Chemical 2-Methoxymethcathinone (hydrochloride), Purity: ≥98%, 50 mg
SDP
Catalog No. 502894169
Encompass_Preferred
A positional isomer of methedrone, having the methoxy group at the 2 rather than the 4 position; intended for forensic and research applications
Catalog No. 50-289-4169 Supplier Cayman Chemical Supplier No. 900118650 MG
A positional isomer of methedrone, having the methoxy group at the 2 rather than the 4 position; intended for forensic and research applications