Learn More
Cayman Chemical 2-Methylmethcathinone (hydrochloride), Purity: ≥98%, CAS: 1246815-51-9, 50 mg
SDP
Catalog No. 502951489
Encompass_Preferred
An analytical reference standard categorized as a cathinone; a potential impurity in the synthesis of 4-methylmethcathinone; intended for research and forensic applications
Catalog No. 50-295-1489 Supplier Cayman Chemical Supplier No. 1122350 MG
An analytical reference standard categorized as a cathinone; a potential impurity in the synthesis of 4-methylmethcathinone; intended for research and forensic applications