Learn More
Vector Laboratories Vicia Villosa Lectin (VVL, VVA) Biotinylated 2 mg
SDP
Catalog No. NC0103175
Encompass
Vicia villosa lectin (VVL, VVA) preferentially binds α- or β-linked terminal N-acetylgalactosamine, especially a single α-N-acetylgalactosamine residue linked to serine or threonine in a polypeptide (the Tn antigen). This lectin may require specific amino acid sequences at the receptor site of glycosylation, and the disaccharide galactosyl (α-1,3) N-acetylgalactosamine is also a potent inhibitor. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC0103175 Supplier Vector Laboratories Supplier No. B12352
Vicia villosa lectin (VVL, VVA) preferentially binds α- or β-linked terminal N-acetylgalactosamine, especially a single α-N-acetylgalactosamine residue linked to serine or threonine in a polypeptide (the Tn antigen). This lectin may require specific amino acid sequences at the receptor site of glycosylation, and the disaccharide galactosyl (α-1,3) N-acetylgalactosamine is also a potent inhibitor. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.