Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration - 2 mL, 0.2 m PTFE membrane (5/pkg)
Supplier:  Cytiva 8247
Encompass
Robust Chemical Resistance Provides Compatibility with Harsh Organics and Aqueous SolutionsChemically resistant materials provide a stable platform to process samples in organic solventsInert materials of construction ensure complete recovery of samples and low non-specific bindingNew well design results in faster, more uniform filtration rates across the plate and reduces hold-up volumeInnovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1368328