Learn More
GENTECH SOLUTIONS LLC 2mL 11.60 x 32 Clear Screw Vial
Supplier:  GENTECH SOLUTIONS LLC 2000100010
Encompass_Preferred
" EliteChem 2ml 9mm Screw Thread Autosampler Vials are designed for high-precision chromatography applications, delivering exceptional durability, chemical resistance, and consistent performance for reliable analysis results.Key Features:Premium USP Type I Borosilicate Glass: Made from 49-Expansion & 33-Expansion USP Type I Borosilicate Glass, known for its exceptional durability, low leachable elements, and superior chemical resistance. The low expansion coefficient ensures minimal thermal stress and consistent performance under varying temperatures.Universal Compatibility: Standard 12x32mm dimensions with 9mm screw thread, fully compatible with leading autosampler brands such as Agilent, Waters, Thermo, Bruker, Varian, and more.Secure 9-425 Screw Thread Design: The precision screw thread design provides a leak-proof, airtight seal, preventing evaporation and ensuring sample integrity. Vials can be reused without compromisi
Catalog No. NC3728803