Learn More
Alkali Scientific 2mL 9 425 Wide Opening PP Screw Vial with Graduations, 100/Pk, Amber
SDP
Supplier:  Alkali Scientific MVP02AMB
Encompass_Preferred
The Alkali Scientific™ 2 mL Wide Opening PP Screw Vial is designed for precise sample handling and reliable storage in laboratory and analytical applications. Made from high quality polypropylene (PP), this vial offers excellent chemical resistance and durability. The wide opening allows for easy filling and cleaning, while the graduated markings provide convenient and accurate volume measurement. Featuring a 9 425 screw thread compatible with standard caps and closures, this vial is ideal for autosampler and routine laboratory workflows. Supplied in packs of 100, it is perfect for chromatography, analytical testing, and small volume sample storage. Key Features 2 mL capacity for precise sample handling and storage Made from durable polypropylene (PP) with excellent chemical resistance Wide opening for easy filling, dispensing, and cleaning Graduated markings for accurate volume measurement 9 425 screw thread compatible with standard caps and closures Ideal for
Catalog No. 50-006-62109