Learn More
Analytical Sales & Services 2mL 96-Well Collection Plate with Round-Bottom Wells, with SiliGuard™ Low Bind Coating, Irradiated
Catalog No. 5000807131
Encompass
Advantage™ 2mL 96-Well Collection Plate with SiliGuard™, Irradiated\n\n• Round Wells with Round Bottoms\n• Polypropylene\n• SiliGuard™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25
Catalog No. 50-008-07131 Supplier Analytical Sales & Services Supplier No. 967720GCRAD
Advantage™ 2mL 96-Well Collection Plate with SiliGuard™, Irradiated\n\n• Round Wells with Round Bottoms\n• Polypropylene\n• SiliGuard™ Low Bind Coating\n• Irradiated\n• Made in USA of domestically sourced and certified materials\n\nQty/Pkg: 25